Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(methylcarbamoyl)benzeneboronic acid, 95%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(methylcarbamoyl)benzeneboronic acid
Formula Weight 196.97
MDL Number MFCD28098710
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5639003

  SDS
alfa aesar™
H5639003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH56390MD

  SDS
alfa aesar™
H56390MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 196.97
Chemical Name or Material 2-Fluoro-3-(methylcarbamoyl)benzeneboronic acid
Physical Form Solid
Assay 95%
MDL Number MFCD28098710
Quantity 1g