Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-methylbenzeneboronic acid pinacol ester, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-methylbenzeneboronic acid pinacol ester
CAS 1192548-08-5
Formula Weight 236.09
MDL Number MFCD18384075
Synonym 2-Fluoro-3-methylphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5639103

 
alfa aesar™
H5639103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH56391MD

 
alfa aesar™
H56391MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 236.09
Chemical Name or Material 2-Fluoro-3-methylbenzeneboronic acid pinacol ester
Physical Form Solid
Assay 97%
MDL Number MFCD18384075
CAS 1192548-08-5
Synonym 2-Fluoro-3-methylphenylboronic acid pinacol ester
Quantity 1g