missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-3-methoxybenzeneboronic acid, 97%, Thermo Scientific Chemicals
$231.71 - $231.71
Chemical Identifiers
CAS | 352303-67-4 |
---|---|
Molecular Formula | C7H8BFO3 |
Molecular Weight (g/mol) | 169.946 |
MDL Number | MFCD02094728 |
InChI Key | JCKZNMSBFBPDPM-UHFFFAOYSA-N |
Synonym | 2-fluoro-3-methoxyphenyl boronic acid, 2-fluoro-3-methoxybenzeneboronic acid, 2-fluoro-3-methoxyphenyl boranediol, 2-fluoro-3-methoxyphenylboronicacid, boronic acid, 2-fluoro-3-methoxyphenyl, pubchem1848, 2-fluoro-3-methoxy-phenyl boronic acid, acmc-1af04, 2-fluoro-3-methoxy-phenylboronic acid |
PubChem CID | 4985744 |
IUPAC Name | (2-fluoro-3-methoxyphenyl)boronic acid |
SMILES | B(C1=C(C(=CC=C1)OC)F)(O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL1965506
|
Thermo Scientific Chemicals
L1965506 |
5 g |
Each for $231.71
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
352303-67-4 | |
169.946 | |
JCKZNMSBFBPDPM-UHFFFAOYSA-N | |
4985744 | |
B(C1=C(C(=CC=C1)OC)F)(O)O |
C7H8BFO3 | |
MFCD02094728 | |
2-fluoro-3-methoxyphenyl boronic acid, 2-fluoro-3-methoxybenzeneboronic acid, 2-fluoro-3-methoxyphenyl boranediol, 2-fluoro-3-methoxyphenylboronicacid, boronic acid, 2-fluoro-3-methoxyphenyl, pubchem1848, 2-fluoro-3-methoxy-phenyl boronic acid, acmc-1af04, 2-fluoro-3-methoxy-phenylboronic acid | |
(2-fluoro-3-methoxyphenyl)boronic acid |
Specifications
352303-67-4 | |
MFCD02094728 | |
9191510 | |
JCKZNMSBFBPDPM-UHFFFAOYSA-N | |
(2-fluoro-3-methoxyphenyl)boronic acid | |
4985744 | |
97% |
C7H8BFO3 | |
5 g | |
2-fluoro-3-methoxyphenyl boronic acid, 2-fluoro-3-methoxybenzeneboronic acid, 2-fluoro-3-methoxyphenyl boranediol, 2-fluoro-3-methoxyphenylboronicacid, boronic acid, 2-fluoro-3-methoxyphenyl, pubchem1848, 2-fluoro-3-methoxy-phenyl boronic acid, acmc-1af04, 2-fluoro-3-methoxy-phenylboronic acid | |
B(C1=C(C(=CC=C1)OC)F)(O)O | |
169.946 | |
169.95 | |
2-Fluoro-3-methoxybenzeneboronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only