Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-methoxybenzaldehyde, 97%

$62.84 - $186.84

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-methoxybenzaldehyde
CAS 103438-88-6
Molecular Formula C8H7FO2
Formula Weight 154.14
MDL Number MFCD03411962
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2672903

  SDS
alfa aesar™
H2672903
1g Each for $62.84

AAH2672906

  SDS
alfa aesar™
H2672906
5g Each for $186.84
Description & Specifications

Specifications

Formula Weight 154.14
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 2-Fluoro-3-methoxybenzaldehyde
Assay 97%
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 50° to 53°C
Quantity 1g
CAS 103438-88-6
MDL Number MFCD03411962
Flash Point >110°C (230°F)
Molecular Formula C8H7FO2
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.