Please login to your online account to display your discounted pricing

2-Fluoro-3-methoxybenzaldehyde, 96%, ACROS Organics™


Quantity 5g

Manufacturer: thermo fisher scientific  AC425830050

 View more versions of this product

Catalog No. AC425830050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 154.14
Quantity 5g
Chemical Name or Material 2-Fluoro-3-methoxybenzaldehyde
Molecular Formula C8H7FO2
MDL Number MFCD03411962
CAS 103438-88-6
Assay Percent Range 96%
Linear Formula FC6H3(OCH3)CHO

Specifications

Chemical Name or Material 2-Fluoro-3-methoxybenzaldehyde
Assay Percent Range 96%
Molecular Formula C8H7FO2
Linear Formula FC6H3(OCH3)CHO
Formula Weight 154.14
MDL Number MFCD03411962