missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Fluoro-3-methoxybenzaldehyde, 96%, Thermo Scientific™
Chemical Identifiers
CAS | 103438-88-6 |
---|---|
Molecular Formula | C8H7FO2 |
Molecular Weight (g/mol) | 154.14 |
MDL Number | MFCD03411962 |
InChI Key | LIHCOUDNHILORI-UHFFFAOYSA-N |
Synonym | 2-fluoro-3-methoxy benzaldehyde, 2-fluoro-3-formylanisole, benzaldehyde, 2-fluoro-3-methoxy, zlchem 868, pubchem4224, acmc-1bs4m, flouro-3-methoxybenzaldehyde, 2-fluoro-3-methoxy-benzaldehyde, benzaldehyde,2-fluoro-3-methoxy, 2-fluoro-3-methoxybenzaldehyde |
PubChem CID | 3774463 |
IUPAC Name | 2-fluoro-3-methoxybenzaldehyde |
SMILES | COC1=CC=CC(C=O)=C1F |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC425830010
|
Thermo Scientific Chemicals
425830010 |
1g |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
103438-88-6 | |
154.14 | |
LIHCOUDNHILORI-UHFFFAOYSA-N | |
3774463 | |
COC1=CC=CC(C=O)=C1F |
C8H7FO2 | |
MFCD03411962 | |
2-fluoro-3-methoxy benzaldehyde, 2-fluoro-3-formylanisole, benzaldehyde, 2-fluoro-3-methoxy, zlchem 868, pubchem4224, acmc-1bs4m, flouro-3-methoxybenzaldehyde, 2-fluoro-3-methoxy-benzaldehyde, benzaldehyde,2-fluoro-3-methoxy, 2-fluoro-3-methoxybenzaldehyde | |
2-fluoro-3-methoxybenzaldehyde |
Specifications
103438-88-6 | |
95% min. (GC) | |
C8H7FO2 | |
MFCD03411962 | |
2-fluoro-3-methoxy benzaldehyde, 2-fluoro-3-formylanisole, benzaldehyde, 2-fluoro-3-methoxy, zlchem 868, pubchem4224, acmc-1bs4m, flouro-3-methoxybenzaldehyde, 2-fluoro-3-methoxy-benzaldehyde, benzaldehyde,2-fluoro-3-methoxy, 2-fluoro-3-methoxybenzaldehyde | |
COC1=CC=CC(C=O)=C1F | |
154.14 | |
154.14 | |
2-Fluoro-3-methoxybenzaldehyde |
>110°C | |
Glass Bottle | |
FC6H3(OCH3)CHO | |
1g | |
LIHCOUDNHILORI-UHFFFAOYSA-N | |
2-fluoro-3-methoxybenzaldehyde | |
3774463 | |
96% |
Safety and Handling
GHS Signal Word: Warning
RUO â Research Use Only