Please login to your online account to display your discounted pricing

2-Fluoro-3-iodopyridine, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-iodopyridine
CAS 113975-22-7
CAS Min % 97.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H3FIN
Formula Weight 222.98
MDL Number MFCD03095287
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

MO00359DA

 
fisher scientific company
MO00359DA
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 222.98
Quantity 1g
Assay 97%
CAS Min % 97.0
Chemical Name or Material 2-Fluoro-3-iodopyridine
Assay Percent Range 97%
Molecular Formula C5H3FIN
MDL Number MFCD03095287
CAS 113975-22-7
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Health Hazard 2 GHS P Statement Causes skin irritation. Causes serious eye irritation.