Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-iodopyridine, 97%

$92.22 - $317.09

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-iodopyridine
CAS 113975-22-7
Molecular Formula C5H3FIN
Formula Weight 222.99
MDL Number MFCD03095287
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5014603

 
alfa aesar™
H5014603
1g Each for $317.09

AAH50146MD

 
alfa aesar™
H50146MD
250mg Each for $92.22
Description & Specifications

Specifications

Formula Weight 222.99
Chemical Name or Material 2-Fluoro-3-iodopyridine
Assay 97%
Molecular Formula C5H3FIN
MDL Number MFCD03095287
CAS 113975-22-7
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.