missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-3-hydroxypyridine 98.0+%, TCI America™

$189.50 - $647.31

Chemical Identifiers

CAS 174669-74-0
Molecular Formula C5H4FNO
Molecular Weight (g/mol) 113.091
MDL Number MFCD04112569
InChI Key UEQRKEWMEMJXQO-UHFFFAOYSA-N
Synonym 2-fluoro-3-hydroxypyridine, 2-fluoro-3-pyridinol, 2-fluoropyridine-3-ol, 3-pyridinol, 2-fluoro, 2-fluoro-3-hydroxy pyridine, pubchem6290, acmc-209e8o, 3-hydroxy-2-fluoropyridine, 3-pyridinol, 2-fluoro-9ci
PubChem CID 2762785
IUPAC Name 2-fluoropyridin-3-ol
SMILES C1=CC(=C(N=C1)F)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
F08861G View Documents Tci America
F08861G
1g Each for $189.50
 
F08865G View Documents Tci America
F08865G
5g Each for $647.31
 
Specifications

Chemical Identifiers

174669-74-0
113.091
UEQRKEWMEMJXQO-UHFFFAOYSA-N
2762785
C1=CC(=C(N=C1)F)O
C5H4FNO
MFCD04112569
2-fluoro-3-hydroxypyridine, 2-fluoro-3-pyridinol, 2-fluoropyridine-3-ol, 3-pyridinol, 2-fluoro, 2-fluoro-3-hydroxy pyridine, pubchem6290, acmc-209e8o, 3-hydroxy-2-fluoropyridine, 3-pyridinol, 2-fluoro-9ci
2-fluoropyridin-3-ol

Specifications

White-Yellow
1g
C5H4FNO
2-fluoro-3-hydroxypyridine, 2-fluoro-3-pyridinol, 2-fluoropyridine-3-ol, 3-pyridinol, 2-fluoro, 2-fluoro-3-hydroxy pyridine, pubchem6290, acmc-209e8o, 3-hydroxy-2-fluoropyridine, 3-pyridinol, 2-fluoro-9ci
C1=CC(=C(N=C1)F)O
113.091
113.09
≥98.0% (GC,T)
128°C
174669-74-0
MFCD04112569
UEQRKEWMEMJXQO-UHFFFAOYSA-N
2-fluoropyridin-3-ol
2762785
Crystal-Powder at 20°C
2-Fluoro-3-hydroxypyridine
Safety and Handling

Safety and Handling

TSCA : No