Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-hydroxypyridine, 97%

$132.07 - $1,226.81

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-hydroxypyridine
CAS 174669-74-0
Molecular Formula C5H4FNO
Formula Weight 113.09
MDL Number MFCD04112569
Synonym 2-Fluoro-3-pyridinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3286203

  SDS
alfa aesar™
H3286203
1g Each for $367.89

AAH3286206

  SDS
alfa aesar™
H3286206
5g Each for $1,226.81

AAH32862MD

  SDS
alfa aesar™
H32862MD
250mg Each for $132.07
Description & Specifications

Specifications

Formula Weight 113.09
Chemical Name or Material 2-Fluoro-3-hydroxypyridine
Assay 97%
Molecular Formula C5H4FNO
MDL Number MFCD04112569
Synonym 2-Fluoro-3-pyridinol
Melting Point 131° to 133°C
Quantity 1g
CAS 174669-74-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.