Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-hydroxy-6-nitrophenylacetone, 97%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-hydroxy-6-nitrophenylacetone
CAS 649736-31-2
Formula Weight 213.16
MDL Number MFCD11869874
Beilstein 11804446
Synonym 1-(2-Fluoro-3-hydroxy-6-nitrophenyl)-2-propanone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6542703

 
alfa aesar™
H6542703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH65427MD

 
alfa aesar™
H65427MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 213.16
Chemical Name or Material 2-Fluoro-3-hydroxy-6-nitrophenylacetone
CAS 649736-31-2
Physical Form Solid
Storage Note 1 2° to 8°C
Beilstein 11804446
Quantity 1g
Synonym 1-(2-Fluoro-3-hydroxy-6-nitrophenyl)-2-propanone
MDL Number MFCD11869874
Assay 97%
Safety and Handling

2° to 8°C