missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-formylbenzeneboronic acid pinacol ester, 96%

Manufacturer:  Alfa Aesar™ H6236203

 View more versions of this product

Catalog No. AAH6236203



Description

Description

Applications
2-Fluoro-3-formylbenzeneboronic acid pinacol ester is used as a intermediate in pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

2-Fluoro-3-formylbenzeneboronic acid pinacol ester
1112209-40-1
MFCD17015836
IFIDGFJKIAHLAQ-UHFFFAOYSA-N
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
53217345
Solid
1g
C13H16BFO3
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C=O)F
250.076
250.07
96%

Chemical Identifiers

1112209-40-1
250.076
IFIDGFJKIAHLAQ-UHFFFAOYSA-N
53217345
B1(OC(C(O1)(C)C)(C)C)C2=C(C(=CC=C2)C=O)F
C13H16BFO3
MFCD17015836
2-fluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde
2-fluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents