Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3-(4-morpholinylmethyl)benzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
Formula Weight 321.19
Synonym 2-Fluoro-3-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6245103

 
alfa aesar™
H6245103
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62451MD

 
alfa aesar™
H62451MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 321.19
Chemical Name or Material 2-Fluoro-3-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
Physical Form Solid
Assay 96%
Synonym 2-Fluoro-3-(4-morpholinylmethyl)benzeneboronic acid pinacol ester
Quantity 1g