Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester, 96%

Chemical Identifiers

Chemical Name or Material 2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester
Formula Weight 266.07
Synonym 2-Fluoro-3,4-methylenedioxyphenylboronic acid pinacol ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6258703

 
alfa aesar™
H6258703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62587MD

 
alfa aesar™
H62587MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 266.07
Assay 96%
Synonym 2-Fluoro-3,4-methylenedioxyphenylboronic acid pinacol ester
Quantity 1g
Chemical Name or Material 2-Fluoro-3,4-methylenedioxybenzeneboronic acid pinacol ester
Physical Form Solid