missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Fluoro-1,4-dimethoxybenzene, 97%

Manufacturer:  Alfa Aesar™ L1350503

 View more versions of this product

Catalog No. AAL1350503



Description

Description

Applications
2-Fluoro-1,4-dimethoxybenzene is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications

Specifications

2-Fluoro-1,4-dimethoxybenzene
23°C to 26°C
82830-49-7
MFCD00042487
1,4-dimethoxy-2-fluorobenzene, 2,5-dimethoxyfluorobenzene, 2,5-dimethoxy-1-fluorobenzene, benzene, 2-fluoro-1,4-dimethoxy, pubchem3050, acmc-209pql, 3-fluoro-1,4-dimethoxybenzene, benzene,2-fluoro-1,4-dimethoxy, 2-fluoranyl-1,4-dimethoxy-benzene, 2-fluorohydroquinone dimethyl ether
COC1=CC(=C(C=C1)OC)F
156.156
156.16
119°C to 121°C (40mmHg)
1g
C8H9FO2
6387735
WNCYZVMZKSOPMU-UHFFFAOYSA-N
2-fluoro-1,4-dimethoxybenzene
145002
97%

Chemical Identifiers

82830-49-7
156.156
WNCYZVMZKSOPMU-UHFFFAOYSA-N
145002
COC1=CC(=C(C=C1)OC)F
C8H9FO2
MFCD00042487
1,4-dimethoxy-2-fluorobenzene, 2,5-dimethoxyfluorobenzene, 2,5-dimethoxy-1-fluorobenzene, benzene, 2-fluoro-1,4-dimethoxy, pubchem3050, acmc-209pql, 3-fluoro-1,4-dimethoxybenzene, benzene,2-fluoro-1,4-dimethoxy, 2-fluoranyl-1,4-dimethoxy-benzene, 2-fluorohydroquinone dimethyl ether
2-fluoro-1,4-dimethoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Documents