Please login to your online account to display your discounted pricing

2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Fluoro-1, 3-dimethylimidazolidinium hexafluorophosphate
CAS 164298-27-5
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C5H10FN2·F6P
Formula Weight 262.11
Synonym 1, 3-Dimethyl-2-fluoroimidazolidinium hexafluorophosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC439470050

 
thermo fisher scientific
AC439470050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC439470250

 
thermo fisher scientific
AC439470250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 262.11
Physical Form Powder or Crystalline Powder
Quantity 5g
Color White
CAS Max % 100.0
CAS Min % 96.0
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Molecular Formula C5H10FN2·F6P
CAS 164298-27-5
Assay Percent Range 97%
Synonym 1, 3-Dimethyl-2-fluoroimidazolidinium hexafluorophosphate
Chemical Name or Material 2-Fluoro-1, 3-dimethylimidazolidinium hexafluorophosphate
Infrared Spectrum Authentic
Elemental Analysis 22.00 to 23.83% C, 10.26 to 11.12% N
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.