missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Fluoro-1,3-dimethylimidazolidinium hexafluorophosphate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC439470050

 View more versions of this product

Catalog No. AC439470050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Fluoro-1, 3-dimethylimidazolidinium hexafluorophosphate
Authentic
170°C to 172°C
164298-27-5
100.0
C5H10FN2·F6P
262.11
Powder or Crystalline Powder
22.00 to 23.83% C, 10.26 to 11.12% N
White
5g
96.0
97%
1, 3-Dimethyl-2-fluoroimidazolidinium hexafluorophosphate, DFIH
262.11
97%

Chemical Identifiers

164298-27-5
C5H10FN2·F6P
262.11
1, 3-Dimethyl-2-fluoroimidazolidinium hexafluorophosphate, DFIH
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents