Please login to your online account to display your discounted pricing

2-Fluoro-1,3-dimethylbenzene, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 2-Fluoro-1, 3-dimethylbenzene
CAS 443-88-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9F
Linear Formula (CH3)2C6H3F
Formula Weight 124.16
MDL Number MFCD00039215
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

CD05584DA

  SDS
fisher scientific company
CD05584DA
Amber Glass Bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material 2-Fluoro-1, 3-dimethylbenzene
Density 0.988g/mL
Quantity 1g
CAS 443-88-9
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Warning
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Amber Glass Bottle
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9F
Linear Formula (CH3)2C6H3F
Formula Weight 124.16
MDL Number MFCD00039215
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.