Please login to your online account to display your discounted pricing

2-Fluoro-1,3-dimethylbenzene, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CD05584DA

 View more versions of this product

Catalog No. CD05584DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 124.16
Quantity 1g
Assay 97%
Density 0.988g/mL
CAS Max % 100.0
Linear Formula (CH3)2C6H3F
Assay Percent Range 97%
Molecular Formula C8H9F
CAS Min % 96.5
Packaging Amber Glass Bottle
Chemical Name or Material 2-Fluoro-1, 3-dimethylbenzene
MDL Number MFCD00039215
CAS 443-88-9
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Fluoro-1, 3-dimethylbenzene
CAS 443-88-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H9F
Linear Formula (CH3)2C6H3F
Formula Weight 124.16
MDL Number MFCD00039215
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapor.