Please login to your online account to display your discounted pricing

2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Maybridge

Chemical Identifiers

Chemical Name or Material 2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H9FO2
Linear Formula FC6H4C6H4CO2H
Formula Weight 216.21
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SEW05200CB

  SDS
fisher scientific company
SEW05200CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 216.21
Quantity 250mg
Assay 95%
Chemical Name or Material 2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid
Assay Percent Range 95%
Molecular Formula C13H9FO2
CAS 365-12-8
Linear Formula FC6H4C6H4CO2H