Please login to your online account to display your discounted pricing

2-Fluoro[1,1-biphenyl]-4-carboxylic acid, 95%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  SEW05200CB

 View more versions of this product

Catalog No. SEW05200CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 216.21
Quantity 250mg
Assay 95%
Assay Percent Range 95%
CAS 365-12-8
Molecular Formula C13H9FO2
Chemical Name or Material 2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid
Linear Formula FC6H4C6H4CO2H

Specifications

Chemical Name or Material 2'-Fluoro[1, 1'-biphenyl]-4-carboxylic acid
Assay 95%
Assay Percent Range 95%
Molecular Formula C13H9FO2
Linear Formula FC6H4C6H4CO2H
Formula Weight 216.21