missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethynylpyridine, 98%, stab. with 0.01% hydroquinone

Manufacturer:  Alfa Aesar™ L1127414

 View more versions of this product

Catalog No. AAL1127414



Description

Description

Applications
2-Ethynylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.

Solubility
Soluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Specifications

2-Ethynylpyridine
Characteristic
25g
1945-84-2
MFCD00041598
2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine, 2-ethynyl pyridine
C#CC1=CC=CC=N1
103.124
103.12
76°C (168°F)
77°C to 78°C (14mmHg)
stabilized with 0.01% hydroquinone
C7H5N
1098904
NHUBNHMFXQNNMV-UHFFFAOYSA-N
2-ethynylpyridine
137268
98%

Chemical Identifiers

1945-84-2
103.124
NHUBNHMFXQNNMV-UHFFFAOYSA-N
137268
C#CC1=CC=CC=N1
C7H5N
MFCD00041598
2-ethynyl pyridine, 2-pyridylacetylene, pyridine, 2-ethynyl, 2-ethynyl-pyridine, pyridine, ethynyl, pyridin-2-yl acetylene, ethynylpyridine, ethynyl pyridine, ortho-ethynylpyridine, 2-ethynyl pyridine
2-ethynylpyridine
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents