missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Ethynylpyridine, 98+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC295670050
Specifications
2-Ethynylpyridine | |
76°C | |
Glass bottle | |
1.02 | |
Brown | |
1945-84-2 | |
100.0 | |
C7H5N | |
2-ethynyl pyridine, 2-ethynyl-pyridine, 2-pyridylacetylene, ethynyl pyridine, ethynylpyridine, ortho-ethynylpyridine, pyridin-2-yl acetylene, pyridine, 2-ethynyl, pyridine, ethynyl | |
NHUBNHMFXQNNMV-UHFFFAOYSA-N | |
2-ethynylpyridine | |
137268 | |
Liquid |
1.0200g/mL | |
Authentic | |
1.5570 to 1.5610 | |
77.0°C to 78.0°C (14.0mmHg) | |
5g | |
98.0 | |
98% min. (GC) | |
MFCD00041598 | |
Solubility in water: soluble in water | |
C#CC1=CC=CC=N1 | |
103.12 | |
103.12 | |
98+% |
Chemical Identifiers
1945-84-2 | |
103.12 | |
NHUBNHMFXQNNMV-UHFFFAOYSA-N | |
137268 | |
C#CC1=CC=CC=N1 |
C7H5N | |
MFCD00041598 | |
2-ethynyl pyridine, 2-ethynyl-pyridine, 2-pyridylacetylene, ethynyl pyridine, ethynylpyridine, ortho-ethynylpyridine, pyridin-2-yl acetylene, pyridine, 2-ethynyl, pyridine, ethynyl | |
2-ethynylpyridine |