missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethynylpyridine, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC295670250

 View more versions of this product

Catalog No. AC295670250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-Ethynylpyridine
76°C
Glass bottle
1.02
Brown
1945-84-2
100.0
C7H5N
2-ethynyl pyridine
NHUBNHMFXQNNMV-UHFFFAOYSA-N
2-ethynylpyridine
137268
Liquid
1.0200g/mL
Authentic
1.5570 to 1.5610
77.0°C to 78.0°C (14.0mmHg)
25g
98.0
98% min. (GC)
MFCD00041598
Solubility in water: soluble in water
C#CC1=CC=CC=N1
103.12
103.12
98+%

Chemical Identifiers

1945-84-2
103.12
NHUBNHMFXQNNMV-UHFFFAOYSA-N
137268
C#CC1=CC=CC=N1
C7H5N
MFCD00041598
2-ethynyl pyridine
2-ethynylpyridine
SDS Documents