missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, TCI America™

Manufacturer:  TCI America E10741G

 View more versions of this product

Catalog No. E10741G



Specifications

Specifications

2-Ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Yellow
347389-74-6
MFCD13182122
Ethynylboronic Acid Pinacol Ester
B1(OC(C(O1)(C)C)(C)C)C#C
152
152.00
≥95.0% (GC)
140°C
1g
C8H13BO2
3272
ZEOXJIIUMORBNN-UHFFFAOYSA-N
2-ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
15891429
Clear Liquid at 20°C

Chemical Identifiers

347389-74-6
152
ZEOXJIIUMORBNN-UHFFFAOYSA-N
15891429
B1(OC(C(O1)(C)C)(C)C)C#C
C8H13BO2
MFCD13182122
Ethynylboronic Acid Pinacol Ester
2-ethynyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Refrigerator

Documents