missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethyltoluene, TCI America™

Manufacturer:  TCI America E01845ML

 View more versions of this product

Catalog No. E01845ML



Specifications

Specifications

2-Ethyltoluene
Colorless
5mL
C9H12
3295
HYFLWBNQFMXCPA-UHFFFAOYSA-N
1-ethyl-2-methylbenzene
11903
120.20
≥99.0% (GC)
166°C
-17°C
611-14-3
MFCD00009257
2-ethyltoluene, o-ethyltoluene, o-methylethylbenzene, 1-methyl-2-ethylbenzene, toluene, o-ethyl, ortho-ethyltoluene, benzene, 1-ethyl-2-methyl, 1,2-methylethylbenzene, o-ethyl methylbenzene, o-ethyl methyl benzene
CCC1=CC=CC=C1C
120.195
CHEBI:34276
Clear Liquid at 20°C

Chemical Identifiers

611-14-3
120.195
HYFLWBNQFMXCPA-UHFFFAOYSA-N
11903
1-ethyl-2-methylbenzene
C9H12
MFCD00009257
2-ethyltoluene, o-ethyltoluene, o-methylethylbenzene, 1-methyl-2-ethylbenzene, toluene, o-ethyl, ortho-ethyltoluene, benzene, 1-ethyl-2-methyl, 1,2-methylethylbenzene, o-ethyl methylbenzene, o-ethyl methyl benzene
CHEBI:34276
CCC1=CC=CC=C1C
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0015

RTECSNumber : XT2500000

TSCA : Yes

Documents