missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylthiophenol, 95%

Manufacturer:  Alfa Aesar™ L1149809

 View more versions of this product

Catalog No. AAL1149809



Specifications

Specifications

2-Ethylthiophenol
Stench
10g
C8H10S
UN2810
2-ethylthiophenol, benzenethiol, 2-ethyl, unii-62gbu45i5p, 2-ethyl thiophenol, o-ethylbenzenethiol, fema no. 3345, ethylbenzenethiol, o, 2-ethylphenylmercaptan, benzenethiol, o-ethyl, o-ethylthiophenol
CCC1=CC=CC=C1S
138.228
138.23
80°C (176°F)
203°C to 205°C
4500-58-7
MFCD00010022
2553288
ABROBCBIIWHVNS-UHFFFAOYSA-N
2-ethylbenzenethiol
3734338
95%

Chemical Identifiers

4500-58-7
138.228
ABROBCBIIWHVNS-UHFFFAOYSA-N
3734338
CCC1=CC=CC=C1S
C8H10S
MFCD00010022
2-ethylthiophenol, benzenethiol, 2-ethyl, unii-62gbu45i5p, 2-ethyl thiophenol, o-ethylbenzenethiol, fema no. 3345, ethylbenzenethiol, o, 2-ethylphenylmercaptan, benzenethiol, o-ethyl, o-ethylthiophenol
2-ethylbenzenethiol
Safety and Handling

Safety and Handling

GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

TSCA : TSCA

Documents