Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Ethylthio)nicotinic acid, 98%

$131.61 - $131.61

Chemical Identifiers

Chemical Name or Material 2-(Ethylthio)nicotinic acid
CAS 27868-76-4
Molecular Formula C8H9NO2S
Formula Weight 183.23
MDL Number MFCD00051680
Beilstein 388961
Synonym 2-(Ethylmercapto)nicotinic acid; 2-(Ethylthio)pyridine-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0780106

 
alfa aesar™
L0780106
5g Each for $131.61
Description & Specifications

Specifications

Formula Weight 183.23
Chemical Name or Material 2-(Ethylthio)nicotinic acid
Assay 98%
Molecular Formula C8H9NO2S
MDL Number MFCD00051680
CAS 27868-76-4
Synonym 2-(Ethylmercapto)nicotinic acid; 2-(Ethylthio)pyridine-3-carboxylic acid
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 5g
Beilstein 388961
Health Hazard 1 Warning
Melting Point 183° to 187°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.