Please login to your online account to display your discounted pricing

2-(Ethylthio)nicotinamide, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  BTB06390EA

 View more versions of this product

Catalog No. BTB06390EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 182.25
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 2-(ethylthio)nicotinamide
CAS 175135-27-0
MDL Number MFCD00052903
CAS Min % 96.5
Packaging Amber Glass Bottle
Assay Percent Range 97%
Molecular Formula C8H10N2OS

Specifications

Chemical Name or Material 2-(ethylthio)nicotinamide
CAS 175135-27-0
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C8H10N2OS
Formula Weight 182.25
MDL Number MFCD00052903