Please login to your online account to display your discounted pricing

2-(Ethylthio)ethylamine +97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-(Ethylthio)ethylamine
CAS 36489-03-9
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C4H11NS
Linear Formula CH3CH2S(CH2)2NH2
Formula Weight 105.2
MDL Number MFCD00014826
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC391260010

  SDS
thermo fisher scientific
AC391260010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.0%
Formula Weight 105.2
Physical Form Liquid
Quantity 1g
Color Clear colorless
Density 0.976g/mL
CAS Min % 97.0
CAS Max % 100.0
Refractive Index 1.4890 to 1.4910 (20°C, 589nm)
Infrared Spectrum Authentic
Chemical Name or Material 2-(Ethylthio)ethylamine
Assay Percent Range 97+%
CAS 36489-03-9
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3CH2S(CH2)2NH2
MDL Number MFCD00014826
Packaging Glass bottle
Molecular Formula C4H11NS
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.