Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Ethylthio)ethylamine, 97%

Chemical Identifiers

Chemical Name or Material 2-(Ethylthio)ethylamine
CAS 36489-03-9
Molecular Formula CH3CH2SCH2CH2NH2
Formula Weight 105.2
MDL Number MFCD00014826
UN Number UN2733
Beilstein 1731375
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2214703

 
alfa aesar™
B2214703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2214706

 
alfa aesar™
B2214706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 105.2
Health Hazard 2 GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
Chemical Name or Material 2-(Ethylthio)ethylamine
Assay 97%
Molecular Formula CH3CH2SCH2CH2NH2
MDL Number MFCD00014826
CAS 36489-03-9
UN Number UN2733
Beilstein 1731375
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Flash Point 52°C (125°F)
Boiling Point 159° to 160°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318-H226
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapour.