missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Ethylthio)ethanol, TCI America™

Manufacturer:  TCI America E0248500ML

 View more versions of this product

Catalog No. E0248500ML



Specifications

Specifications

2-(Ethylthio)ethanol
Yellow
500mL
C4H10OS
1760
LNRIEBFNWGMXKP-UHFFFAOYSA-N
2-ethylsulfanylethanol
8075
Clear Liquid at 20°C
182°C
-100°C
110-77-0
MFCD00002909
2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol
CCSCCO
106.183
106.18
≥98.0% (GC)

Chemical Identifiers

110-77-0
106.183
LNRIEBFNWGMXKP-UHFFFAOYSA-N
8075
CCSCCO
C4H10OS
MFCD00002909
2-ethylthio ethanol, ethyl 2-hydroxyethyl sulfide, ethanol, 2-ethylthio, ethyl thioethanol, 2-hydroxyethyl ethyl sulfide, ethyl 2-hydroxyethyl thioether, beta-ethylthioethanol, 2-ethylsulfanyl ethanol, beta-hydroxydiethyl sulfide, 2-ethylthioethanol
2-ethylsulfanylethanol
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2452

RTECSNumber : KL1225000

TSCA : Yes

Documents