Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Ethylthio)ethanol, 96%

Chemical Identifiers

Chemical Name or Material 2-(Ethylthio)ethanol
CAS 110-77-0
Molecular Formula CH3CH2SCH2CH2OH
Formula Weight 106.19
MDL Number MFCD00002909
UN Number UN1760
Beilstein 1731373
Merck Index 14,3857
Synonym Ethyl 2-hydroxyethyl sulfide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0504018

  SDS
alfa aesar™
L0504018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 106.19
CAS 110-77-0
Molecular Formula CH3CH2SCH2CH2OH
Chemical Name or Material 2-(Ethylthio)ethanol
Assay 96%
MDL Number MFCD00002909
Quantity 50g
RTECS Number KL1225000
Synonym Ethyl 2-hydroxyethyl sulfide
TSCA TSCA
Health Hazard 1 Danger
Flash Point 93°C (199°F)
Boiling Point 180° to 184°C
Melting Point ≈−100°C
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
Health Hazard 2 GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
UN Number UN1760
Beilstein 1731373
Merck Index 14,3857
Safety and Handling

Danger

GHS P Statement
P261-P280-P305+P351+P338-P309-P310
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.