Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Ethylsulfonyl)ethanol, 94%

$227.81 - $227.81

Chemical Identifiers

Chemical Name or Material 2-(Ethylsulfonyl)ethanol
CAS 513-12-2
Molecular Formula C4H10O3S
Formula Weight 138.19
MDL Number MFCD00041259
Beilstein 1748754
Synonym 2-(Ethanesulfonyl)ethanol; Ethyl 2-hydroxyethyl sulfone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2018014

  SDS
alfa aesar™
B2018014
25g Each for $227.81
Description & Specifications

Specifications

Formula Weight 138.19
Chemical Name or Material 2-(Ethylsulfonyl)ethanol
Assay 94%
Molecular Formula C4H10O3S
MDL Number MFCD00041259
CAS 513-12-2
Beilstein 1748754
Synonym 2-(Ethanesulfonyl)ethanol; Ethyl 2-hydroxyethyl sulfone
Boiling Point 153°C (2.5mmHg)
Melting Point 35° to 41°C
Quantity 25g
Flash Point 188°C (370°F)
RTECS Number KL1100000