Please login to your online account to display your discounted pricing

2-Ethylpyrrole, 98%, ACROS Organics™


Quantity 250mg
Packaging Glass Ampule with screw-cap

Manufacturer: thermo fisher scientific  AC308772500

 View more versions of this product

Catalog No. AC308772500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Assay ≥97.5%
Formula Weight 95.15
Quantity 250mg
Color Clear yellow to brown
Density 0.929g/mL
CAS Max % 100.0
Chemical Name or Material 2-Ethylpyrrole
Assay Percent Range 98%
Molecular Formula C6H9N
MDL Number MFCD00060477
CAS 1551-06-0
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass Ampule with screw-cap
Refractive Index 1.4985 to 1.5025 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Ethylpyrrole
CAS 1551-06-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H9N
Formula Weight 95.15
MDL Number MFCD00060477
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents