missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylpyridine, TCI America™

Manufacturer:  TCI America E0168100ML

 View more versions of this product

Catalog No. E0168100ML



Specifications

Specifications

2-Ethylpyridine
Yellow
100mL
C7H9N
1993
NRGGMCIBEHEAIL-UHFFFAOYSA-N
2-ethylpyridine
7523
Clear Liquid at 20°C
144°C
-63°C
100-71-0
MFCD00006361
pyridine, 2-ethyl, ethylpyridine, 2-ethyl-pyridine, pyridine, ethyl, alpha-ethylpyridine, .alpha.-ethylpyridine, unii-06x1w46pyx, 2-ethylpyridin, 2-ethylpyridine, acmc-2097ry
CCC1=CC=CC=N1
107.156
107.16
≥98.0% (GC,T)

Chemical Identifiers

100-71-0
107.156
NRGGMCIBEHEAIL-UHFFFAOYSA-N
7523
CCC1=CC=CC=N1
C7H9N
MFCD00006361
pyridine, 2-ethyl, ethylpyridine, 2-ethyl-pyridine, pyridine, ethyl, alpha-ethylpyridine, .alpha.-ethylpyridine, unii-06x1w46pyx, 2-ethylpyridin, 2-ethylpyridine, acmc-2097ry
2-ethylpyridine
Safety and Handling

Safety and Handling

EINECSNumber : (9)-0222

TSCA : Yes

Documents