Please login to your online account to display your discounted pricing

2-Ethylpyridine, 98+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Ethylpyridine
Name Note 98+%
CAS 100-71-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C7H9N
Linear Formula (C5H4N)CH2CH3
Formula Weight 107.15
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC150565000

  SDS
thermo fisher scientific
150565000
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Liquid
Formula Weight 107.15
Quantity 500g
Color Clear colorless to yellow
TSCA TSCA
EINECS Number 202-881-9
Density 0.937g/mL
Beilstein 20, 241
CAS Min % 98.0
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.4960 to 1.4980 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning
Chemical Name or Material 2-Ethylpyridine
Linear Formula (C5H4N)CH2CH3
CAS 100-71-0
Assay Percent Range 98+%
Name Note 98+%
Molecular Formula C7H9N
MDL Number MFCD00006361
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Flammable liquid and vapor.