Learn More
2-Ethylpyridine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1735222
Description
2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Ethylpyridine is used to study its effect on the proliferation and survival of human umbilical vein endothelial cells (HUVECs), HMVECs from lung, and NIH 3T3 cells. 2-Ethylpyridine linked with silica is a popular stationary phase for chiral and achiral separations in supercritical fluid chromatography.
Solubility
Miscible with water (42 g/L at20°C).
Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications
2-Ethylpyridine | |
-63°C | |
9 to 10 | |
37°C (98°F) | |
C7H9N | |
MFCD00006361 | |
UN1993 | |
pyridine, 2-ethyl, ethylpyridine, 2-ethyl-pyridine, pyridine, ethyl, alpha-ethylpyridine, .alpha.-ethylpyridine, unii-06x1w46pyx, 2-ethylpyridin, 2-ethylpyridine, acmc-2097ry | |
NRGGMCIBEHEAIL-UHFFFAOYSA-N | |
2-ethylpyridine | |
7523 | |
98% |
100-71-0 | |
0.937 | |
149°C | |
Unpleasant | |
1.497 | |
100 g | |
106480 | |
Miscible with water (42g/L at20°C). | |
CCC1=CC=CC=N1 | |
107.156 | |
107.16 |
Chemical Identifiers
100-71-0 | |
107.156 | |
NRGGMCIBEHEAIL-UHFFFAOYSA-N | |
7523 | |
CCC1=CC=CC=N1 |
C7H9N | |
MFCD00006361 | |
pyridine, 2-ethyl, ethylpyridine, 2-ethyl-pyridine, pyridine, ethyl, alpha-ethylpyridine, .alpha.-ethylpyridine, unii-06x1w46pyx, 2-ethylpyridin, 2-ethylpyridine, acmc-2097ry | |
2-ethylpyridine |
Safety and Handling
GHS H Statement
H226-H302-H312-H332-H315-H319-H335
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H315-H319-H332
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 202-881-9
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only