Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylpyrazine, 99%

Chemical Identifiers

Chemical Name or Material 2-Ethylpyrazine
CAS 13925-00-3
Molecular Formula C6H8N2
Formula Weight 108.14
MDL Number MFCD00006149
UN Number UN1993
Beilstein 108200
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1275906

  SDS
alfa aesar™
A1275906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 108.14
Chemical Name or Material 2-Ethylpyrazine
TSCA TSCA
Assay 99%
Molecular Formula C6H8N2
MDL Number MFCD00006149
CAS 13925-00-3
UN Number UN1993
Beilstein 108200
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
RTECS Number UQ3330000
Flash Point 42°C (107°F)
Boiling Point 152° to 153°C
Quantity 5g
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.