Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylphenyl isothiocyanate, 98%


Quantity 25g

Manufacturer: alfa aesar™  L0982114

 View more versions of this product

Catalog No. AAL0982114

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 163.24
Chemical Name or Material 2-Ethylphenyl isothiocyanate
Molecular Formula C9H9NS
CAS 19241-19-1
Health Hazard 2 GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00041066
UN Number UN2922
Beilstein 2802909
Health Hazard 1 Danger
Boiling Point 124° to 125°C (15mmHg)
Quantity 25g
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Assay 98%
Odor Pungent
Flash Point >110°C (230°F)

Specifications

Chemical Name or Material 2-Ethylphenyl isothiocyanate
CAS 19241-19-1
Molecular Formula C9H9NS
Formula Weight 163.24
MDL Number MFCD00041066
UN Number UN2922
Beilstein 2802909
Accessories

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.