Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylphenyl isocyanate, 99%

Chemical Identifiers

Chemical Name or Material 2-Ethylphenyl isocyanate
CAS 40411-25-4
Molecular Formula C9H9NO
Formula Weight 147.18
MDL Number MFCD00002013
UN Number UN2206
Beilstein 3538910
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1525206

 
alfa aesar™
L1525206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 147.18
Molecular Formula C9H9NO
MDL Number MFCD00002013
Chemical Name or Material 2-Ethylphenyl isocyanate
Assay 99%
CAS 40411-25-4
Beilstein 3538910
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 56°C (3mmHg)
Quantity 5g
UN Number UN2206
Recommended Storage 2° to 8°C
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: ISOCYANATES, TOXIC, N.O.S.
Flash Point 78°C (172°F)
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H330-H334-H227-H315-H319-H335
Fatal if swallowed.
Fatal if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C