missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylnitrobenzene, TCI America™

Manufacturer:  TCI America E015325G

 View more versions of this product

Catalog No. E015325G



Specifications

Specifications

2-Ethylnitrobenzene
Yellow
25g
C8H9NO2
2-ethylnitrobenzene, o-ethylnitrobenzene, benzene, 1-ethyl-2-nitro, ethylnitrobenzene, o-nitroethylbenzene, 2-ethyl-1-nitrobenzene, 1-ethy1-2-nitro-benzene, ccris 3099, o-ethyl nitrobenzene, benzene, ethylnitro
CCC1=CC=CC=C1[N+](=O)[O-]
151.165
151.17
≥99.0% (GC)
232°C
-13°C
612-22-6
MFCD00024294
PXWYZLWEKCMTEZ-UHFFFAOYSA-N
1-ethyl-2-nitrobenzene
69156
Clear Liquid at 20°C

Chemical Identifiers

612-22-6
151.165
PXWYZLWEKCMTEZ-UHFFFAOYSA-N
69156
CCC1=CC=CC=C1[N+](=O)[O-]
C8H9NO2
MFCD00024294
2-ethylnitrobenzene, o-ethylnitrobenzene, benzene, 1-ethyl-2-nitro, ethylnitrobenzene, o-nitroethylbenzene, 2-ethyl-1-nitrobenzene, 1-ethy1-2-nitro-benzene, ccris 3099, o-ethyl nitrobenzene, benzene, ethylnitro
1-ethyl-2-nitrobenzene
Safety and Handling

Safety and Handling

EINECSNumber : (9)-2453

TSCA : No

Documents