Learn More
2-Ethylhexylamine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC271661000
Specifications
2-Ethylhexylamine | |
50°C | |
Glass bottle | |
0.78 | |
169.0°C | |
-76.0°C | |
104-75-6 | |
100.0 | |
C8H19N | |
MFCD00008148 | |
1-amino-2-ethylhexan, 1-amino-2-ethylhexane, 1-hexanamine, 2-ethyl, 2-ethyl hexylamine, 2-ethyl-1-hexylamine, 2-ethylhexanamine, 2-ethylhexylamine, beta-ethylhexylamine, hexylamine, 2-ethyl, isooctylame | |
LTHNHFOGQMKPOV-UHFFFAOYSA-N | |
2-ethylhexan-1-amine | |
7719 | |
Liquid |
0.7800g/mL | |
Authentic | |
1.4295 to 1.4315 | |
1.1 mPa.s (20°C) | |
Colorless | |
100mL | |
98.5 | |
99% | |
CH3(CH2)3CH(C2H5)CH2NH2 | |
04,388 | |
Solubility in water: 2.5g/L (20 c). | |
CCCCC(CC)CN | |
129.25 | |
129.25 | |
99% |
Chemical Identifiers
104-75-6 | |
129.25 | |
LTHNHFOGQMKPOV-UHFFFAOYSA-N | |
7719 | |
CCCCC(CC)CN |
C8H19N | |
MFCD00008148 | |
1-amino-2-ethylhexan, 1-amino-2-ethylhexane, 1-hexanamine, 2-ethyl, 2-ethyl hexylamine, 2-ethyl-1-hexylamine, 2-ethylhexanamine, 2-ethylhexylamine, beta-ethylhexylamine, hexylamine, 2-ethyl, isooctylame | |
2-ethylhexan-1-amine |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic if inhaled.
Flammable liquid and vapor.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-233-8