Learn More
Alfa Aesar™ 2-Ethylhexylamine, 98%
$20.19 - $212.81
Chemical Identifiers
CAS | 104-75-6 |
---|---|
Molecular Formula | C8H19N |
Molecular Weight (g/mol) | 129.247 |
MDL Number | MFCD00008148 |
InChI Key | LTHNHFOGQMKPOV-UHFFFAOYSA-N |
Synonym | 2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame |
PubChem CID | 7719 |
IUPAC Name | 2-ethylhexan-1-amine |
SMILES | CCCCC(CC)CN |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA10372AE |
Alfa Aesar™
A10372AE |
100mL |
Each for $20.19
|
|
AAA10372AP |
Alfa Aesar™
A10372AP |
500mL |
Each for $53.82
|
|
AAA103720F |
Alfa Aesar™
A103720F |
2500mL |
Each for $212.81
|
|
Description
Applications2-Ethylhexylamine is used to make pharmaceuticals and other chemicals. It is used as beam material and surface active agent. It also involves in synthesis of aliphatic amines.
Solubility
Soluble in water. 2.5 g/L (20°C)
Notes
Air Sensitive. Incompatible with strong oxidizing agents. Protect against electrostatic charges. Store away from acid chlorides and acid anhydrides.
Chemical Identifiers
104-75-6 | |
129.247 | |
LTHNHFOGQMKPOV-UHFFFAOYSA-N | |
7719 | |
CCCCC(CC)CN |
C8H19N | |
MFCD00008148 | |
2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame | |
2-ethylhexan-1-amine |
Specifications
50°C (122°F) | |
169°C | |
100mL | |
C8H19N | |
UN2276 | |
2-ethylhexylamine, 1-hexanamine, 2-ethyl, 2-ethyl-1-hexylamine, 2-ethyl hexylamine, 2-ethylhexanamine, 1-amino-2-ethylhexane, hexylamine, 2-ethyl, beta-ethylhexylamine, 1-amino-2-ethylhexan, isooctylame | |
CCCCC(CC)CN | |
129.247 | |
129.25 | |
2-Ethylhexylamine |
Amine-like | |
-76°C | |
104-75-6 | |
MFCD00008148 | |
1209249 | |
LTHNHFOGQMKPOV-UHFFFAOYSA-N | |
2-ethylhexan-1-amine | |
7719 | |
98% |
Safety and Handling
GHS H Statement
H311-H314-H318-H226-H302
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
GHS P Statement
P210-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: 2-ETHYLHEXYLAMINE
RTECSNumber : MQ5250000
TSCA : TSCA