Please login to your online account to display your discounted pricing

2-Ethylhexyl bromide, 95%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: fisher scientific acros  305331000

 View more versions of this product

Catalog No. AC305331000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 193.13
Physical Form Liquid
Quantity 100g
Color Clear colorless to yellow
EINECS Number 242-659-9
TSCA TSCA
Density 1.0800g/mL
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.4530 to 1.4560 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Infrared Spectrum Authentic
Synonym 1-Bromo-2-ethylhexane
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula CH3(CH2)3CH(C2H5)CH2Br
MDL Number MFCD00000220
Assay Percent Range 95%
Name Note 95%
Chemical Name or Material 2-Ethylhexyl bromide
CAS 18908-66-2
Molecular Formula C8H17Br

Specifications

Chemical Name or Material 2-Ethylhexyl bromide
Name Note 95%
CAS 18908-66-2
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H17Br
Linear Formula CH3(CH2)3CH(C2H5)CH2Br
Formula Weight 193.13
MDL Number MFCD00000220
Synonym 1-Bromo-2-ethylhexane
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents