missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylhexanoyl Chloride, TCI America™

Manufacturer:  TCI America E0336500ML

 View more versions of this product

Catalog No. E0336500ML



Specifications

Specifications

2-Ethylhexanoyl Chloride
Yellow
760-67-8
MFCD00000718
hexanoyl chloride, 2-ethyl, 2-ethylcaproyl chloride, 2-ethylhexanoic acid chloride, 2-ethylhexanoylchlorid, 2-ethylhexanoic acid, chloride, 2-ethyl-hexanoyl chloride, dsstox_cid_7308, dsstox_rid_78399, dsstox_gsid_27308, 2-ethylhexanoylchloride
CCCCC(CC)C(=O)Cl
162.657
162.66
≥98.0% (GC,T)
101°C
500mL
C8H15ClO
3265
WFSGQBNCVASPMW-UHFFFAOYSA-N
2-ethylhexanoyl chloride
79103
Clear Liquid at 20°C

Chemical Identifiers

760-67-8
162.657
WFSGQBNCVASPMW-UHFFFAOYSA-N
79103
CCCCC(CC)C(=O)Cl
C8H15ClO
MFCD00000718
hexanoyl chloride, 2-ethyl, 2-ethylcaproyl chloride, 2-ethylhexanoic acid chloride, 2-ethylhexanoylchlorid, 2-ethylhexanoic acid, chloride, 2-ethyl-hexanoyl chloride, dsstox_cid_7308, dsstox_rid_78399, dsstox_gsid_27308, 2-ethylhexanoylchloride
2-ethylhexanoyl chloride
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0632

RTECSNumber : MP4525000

TSCA : Yes

Documents