Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylhexanoyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 2-Ethylhexanoyl chloride
CAS 760-67-8
Molecular Formula C8H15ClO
Formula Weight 162.66
MDL Number MFCD00000718
UN Number UN2927
Beilstein 956601
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0318114

  SDS
alfa aesar™
L0318114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0318122

  SDS
alfa aesar™
L0318122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0318136

  SDS
alfa aesar™
L0318136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 162.66
MDL Number MFCD00000718
Health Hazard 3 GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Chemical Name or Material 2-Ethylhexanoyl chloride
Odor Pungent
Assay 97%
Molecular Formula C8H15ClO
CAS 760-67-8
UN Number UN2927
Health Hazard 1 Danger
Quantity 25g
Flash Point 79°C (174°F)
Health Hazard 2 GHS H Statement
H330-H314-H318-H227-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC LIQUIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA TSCA
Beilstein 956601
Boiling Point 184°C
Melting Point <−75°C
RTECS Number MP4525000

Applications
2-Ethylhexanoyl chloride was used in the preparation of 4-aza-6-ethyl-4-(2-ethylhexanoyloxy)-5-oxodecyl 2,3, 4,6-tetra-O-acetyl-l-thio-β-D glucopyranoside. It was also used in the preparation of poly(4,8-bis-alkyloxybenzo(1,2-b:4,5-b′)dithiophene-2,6-diyl-alt-(alkyl thieno(3,4-b)thiophene-2-carboxylate)-2,6-diyl), having very promising photovoltaic properties.

Solubility
Reacts with water.

Notes
Moisture Sensitive.Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, moisture, active metals,water, bases.

Safety and Handling

Danger

GHS P Statement
P280-P303+P361+P353-P305+P351+P338-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H330-H314-H318-H227-H302
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Harmful if swallowed.