Learn More
2-Ethylhexanoic acid, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 118520011
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Ethylhexanoic acid | |
98.5 | |
-59.0°C | |
228.0°C | |
Authentic | |
Glass Bottle | |
1.4240 to 1.4260 | |
MFCD00002675 | |
02,349 | |
2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid | |
OBETXYAYXDNJHR-UHFFFAOYSA-N | |
2-ethylhexanoic acid | |
8697 | |
144.21 | |
Liquid |
149-57-5 | |
100.0 | |
0.9060g/mL | |
114°C | |
98.5% min. (GC) | |
C8H16O2 | |
CH3(CH2)3CH(C2H5)CO2H | |
1 kg | |
0.906 | |
Solubility in water: 2g/L (20°C). Other solubilities: soluble in ether | |
CCCCC(CC)C(=O)O | |
144.21 | |
7.7 mPa.s (20°C) | |
99% |
Chemical Identifiers
149-57-5 | |
OBETXYAYXDNJHR-UHFFFAOYSA-N | |
8697 | |
CCCCC(CC)C(=O)O |
MFCD00002675 | |
2-ethylcaproic acid, hexanoic acid, 2-ethyl, ethylhexanoic acid, ethylhexoic acid, 2-ethylhexoic acid, butylethylacetic acid, 2-butylbutanoic acid, 3-heptanecarboxylic acid, ethyl hexanoic acid, 2-ethyl-hexoic acid | |
2-ethylhexanoic acid |
Safety and Handling
GHS H Statement:
Suspected of damaging the unborn child.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 205-743-6
RUO – Research Use Only