Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Ethylfuran, 98+%

Chemical Identifiers

Chemical Name or Material 2-Ethylfuran
CAS 3208-16-0
Molecular Formula C6H8O
Formula Weight 96.13
MDL Number MFCD00003259
UN Number UN3271
Beilstein 105401
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1552006

  SDS
alfa aesar™
A1552006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1552014

  SDS
alfa aesar™
A1552014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1552022

  SDS
alfa aesar™
A1552022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 96.13
UN Number UN3271
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay ≥98%
CAS 3208-16-0
Quantity 5g
Flash Point −2°C (28°F)
Chemical Name or Material 2-Ethylfuran
Boiling Point 92° to 93°C
TSCA TSCA
Beilstein 105401
Molecular Formula C6H8O
MDL Number MFCD00003259
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H225
Highly flammable liquid and vapour.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ETHERS, N.O.S.
Safety and Handling

Danger

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225
Highly flammable liquid and vapour.