missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Ethylcyclohexanol (cis- and trans- mixture), TCI America™

Manufacturer:  TCI America E036525ML

 View more versions of this product

Catalog No. E036525ML



Specifications

Specifications

2-Ethylcyclohexanol (cis- and trans- mixture)
Colorless
3760-20-1
MFCD00001445
CCC1CCCCC1O
128.215
128.22
≥97.0% (GC)
79°C
25mL
C8H16O
CFYUBZHJDXXXQE-UHFFFAOYSA-N
2-ethylcyclohexan-1-ol
19576
Clear Liquid at 20°C

Chemical Identifiers

3760-20-1
128.215
CFYUBZHJDXXXQE-UHFFFAOYSA-N
2-ethylcyclohexan-1-ol
C8H16O
MFCD00001445
19576
CCC1CCCCC1O
Safety and Handling

Safety and Handling

TSCA : No

Documents